Learn More
Medchemexpress LLC (+)-Biotin 4-nitrophenyl ester | 33755-53-2 | ≥98.0% | 500 MG
SDP
Catalog No. 5000340579
Encompass_Preferred
(+)-Biotin-ONP is an alkyl chain-based PROTAC linker that can be utilized in the synthesis of PROTACs. This mechanism leverages the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PROTACs consist of two distinct ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase.
  • The other ligand targets the protein intended for degradation.
Catalog No. 50-003-40579 Supplier Medchemexpress LLC Supplier No. HY130888500MG
(+)-Biotin-ONP is an alkyl chain-based PROTAC linker that can be utilized in the synthesis of PROTACs. This mechanism leverages the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PROTACs consist of two distinct ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase.
  • The other ligand targets the protein intended for degradation.