Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Biotin 4-Nitrophenyl Ester 98.0+%, TCI America™
SDP

Catalog No. B4009200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4009200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4009200MG Supplier TCI America Supplier No. B4009200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33755-53-2
Molecular Formula C16H19N3O5S
Molecular Weight (g/mol) 365.40
MDL Number MFCD15141972
InChI Key YUDNXDTXQPYKCA-YDHLFZDLSA-N
Synonym +-biotin 4-nitrophenyl ester, d-biotin p-nitrophenyl ester, biotinyl-4-nitrophenyl ester, bnp, d-biotin 4-nitrophenyl ester, biotin-p-nitrophenyl ester, 4-nitrophenyl 5-3as,4s,6ar-2-oxo-hexahydrothieno 3,4-d imidazol-4-yl pentanoate, biotin p-nitrophenyl ester, 4-nitrophenyl +-biotinate, +-biotin p-nitrophenyl ester, 4-nitrophenyl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate
PubChem CID 83848
IUPAC Name 4-nitrophenyl 5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoate
SMILES [O-][N+](=O)C1=CC=C(OC(=O)CCCC[C@@H]2SC[C@@H]3NC(=O)N[C@H]23)C=C1

Specifications

Melting Point 165°C
Color White
Quantity 200 mg
Formula Weight 365.40
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Biotin 4-Nitrophenyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.