Learn More
Selleck Chemical LLC Biotin (Vitamin B7) S3130-500mg
SDP
Catalog No. 503110713
Encompass_Preferred
Biotin(Vitamin B7) is a B-vitamin and acts as a necessary coenzyme for cell growth the production of fatty acids and the metabolism of fats and amino acids Biotin is covalently attached to distinct lysine residues in histones affecting chromatin structure and mediating gene regulation
Catalog No. 50-311-0713 Supplier Selleck Chemical LLC Supplier No. S3130500MG
Biotin(Vitamin B7) is a B-vitamin and acts as a necessary coenzyme for cell growth the production of fatty acids and the metabolism of fats and amino acids Biotin is covalently attached to distinct lysine residues in histones affecting chromatin structure and mediating gene regulation