Learn More
Cayman Chemical BIotIn 4AmIdobenzoIc AcI 25mg
SDP
Catalog No. 502982280
Encompass_Preferred
A biotinidase substrate; enzymatic hydrolysis releases PABA which can be quantified by colorimetric detection at 540 nm in the presence of N-1-naphthylethylenediamine as a measure of biotinidase activity; has been used in the evaluation of biotinidase deficiency
Catalog No. 50-298-2280 Supplier Cayman Chemical Supplier No. 2053425 MG
A biotinidase substrate; enzymatic hydrolysis releases PABA which can be quantified by colorimetric detection at 540 nm in the presence of N-1-naphthylethylenediamine as a measure of biotinidase activity; has been used in the evaluation of biotinidase deficiency