Learn More
Cayman Chemical (+)-Biotin 4-Amidobenzoic Acid (sodium salt), Purity: ≥98%, CAS: 102418-74-6, 50 mg
SDP
Catalog No. 502984617
Encompass_Preferred
A biotinidase substrate; enzymatic hydrolysis releases PABA which can be quantified by colorimetric detection at 540 nm in the presence of N-1-naphthylethylenediamine as a measure of biotinidase activity; has been used in the evaluation of biotinidase deficiency
Catalog No. 50-298-4617 Supplier Cayman Chemical Supplier No. 2053450 MG
A biotinidase substrate; enzymatic hydrolysis releases PABA which can be quantified by colorimetric detection at 540 nm in the presence of N-1-naphthylethylenediamine as a measure of biotinidase activity; has been used in the evaluation of biotinidase deficiency