Learn More
AdipoGen Biotin-5-fluorescein-conjugate
SDP
Catalog No. 502561685
Encompass_Preferred
Chemical. CAS 957494-27-8. Formula C33H32N4O8S. MW 644.7. Synthetic. Biotin-fluorescein conjugate for the fluorometric accurate measurement and quantification of avidin and streptavidin. Shows exceptionally fast and tight binding to avidin and streptavidin accompanied by strong quenching. Used in spectrophotometric streptavidin-biotin binding assays. May be used in situations similar to fluorescein isothiocyanate staining to label avidin/streptavidin conjugated molecules such as antibodies with the fluorophore 5-fluorescein. Spectral Data lambdaex 490nm lambdaem 524nm in 0.1M Tris pH8.0.
Catalog No. 50-256-1685 Supplier AdipoGen Supplier No. CDXB0078M005
Chemical. CAS 957494-27-8. Formula C33H32N4O8S. MW 644.7. Synthetic. Biotin-fluorescein conjugate for the fluorometric accurate measurement and quantification of avidin and streptavidin. Shows exceptionally fast and tight binding to avidin and streptavidin accompanied by strong quenching. Used in spectrophotometric streptavidin-biotin binding assays. May be used in situations similar to fluorescein isothiocyanate staining to label avidin/streptavidin conjugated molecules such as antibodies with the fluorophore 5-fluorescein. Spectral Data lambdaex 490nm lambdaem 524nm in 0.1M Tris pH8.0.