Learn More
Medchemexpress LLC HY-B0511 500mg , Biotin CAS:58-85-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0511500MG
Encompass_Preferred
HY-B0511 500mg Biotin CAS: 58-85-5 Biotin (Vitamin B7) is a water-soluble B vitamin and serves as a coenzyme for five carboxylases in humans, involved in the synthesis of fatty acids, isoleucine, and valine, and in gluconeogenesis. Biotin is necessary for cell growth, the production of fatty acids, and the metabolism of fats and amino acids. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9813