Learn More
Medchemexpress LLC Biotin-Ahx-angiotensin II human | 115102-72-2 | 100.0% | 1385.63 g/mol | C66H96N16O15S | 5 MG
SDP
Catalog No. 5000223121
Encompass_Preferred
Biotin-Ahx-Angiotensin II human is a biotin-labeled peptide based on the human angiotensin II sequence, modified with an aminohexanoic acid spacer and provided as a lyophilized powder for research use. It is designed for biochemical and receptor-binding studies where a biotin tag enables affinity capture or detection using streptavidin-based methods.
  • Biotin tag enables streptavidin-based capture and detection.
  • Aminohexanoic acid spacer helps preserve biological activity and accessibility.
  • Sequence corresponds to human angiotensin II for receptor-binding studies.
  • Supplied as a powder for convenient storage and handling.
  • High purity suitable for biochemical and binding assays.
Catalog No. 50-002-23121 Supplier Medchemexpress LLC Supplier No. HY13948F5MG
Biotin-Ahx-Angiotensin II human is a biotin-labeled peptide based on the human angiotensin II sequence, modified with an aminohexanoic acid spacer and provided as a lyophilized powder for research use. It is designed for biochemical and receptor-binding studies where a biotin tag enables affinity capture or detection using streptavidin-based methods.
  • Biotin tag enables streptavidin-based capture and detection.
  • Aminohexanoic acid spacer helps preserve biological activity and accessibility.
  • Sequence corresponds to human angiotensin II for receptor-binding studies.
  • Supplied as a powder for convenient storage and handling.
  • High purity suitable for biochemical and binding assays.