Learn More
Medchemexpress LLC Biotin-Ahx-angiotensin II (human) | 115102-72-2 | 100.0% | C66H96N16O15S | 10MG
SDP
Catalog No. 5000261080
Encompass_Preferred
Biotin-Ahx-Angiotensin II (human) is a biotinylated octapeptide derived from angiotensin II, provided as a lyophilized powder for use as an affinity probe in biochemical and receptor-binding assays. The biotin tag attached via an aminohexanoic acid (Ahx) spacer enables streptavidin-based capture or detection while maintaining the peptide's biological activity. Molecular formula C66H96N16O15S; molecular weight 1385.63.
  • Biotin tag for streptavidin-based detection and capture.
  • Ahx spacer reduces steric hindrance and preserves peptide activity.
  • Sequence corresponds to human angiotensin II for biologically relevant assays.
  • High purity for consistent and reproducible results.
  • Lyophilized powder format for long-term storage and easy reconstitution.
Catalog No. 50-002-61080 Supplier Medchemexpress LLC Supplier No. HY13948F10MG
Biotin-Ahx-Angiotensin II (human) is a biotinylated octapeptide derived from angiotensin II, provided as a lyophilized powder for use as an affinity probe in biochemical and receptor-binding assays. The biotin tag attached via an aminohexanoic acid (Ahx) spacer enables streptavidin-based capture or detection while maintaining the peptide's biological activity. Molecular formula C66H96N16O15S; molecular weight 1385.63.
  • Biotin tag for streptavidin-based detection and capture.
  • Ahx spacer reduces steric hindrance and preserves peptide activity.
  • Sequence corresponds to human angiotensin II for biologically relevant assays.
  • High purity for consistent and reproducible results.
  • Lyophilized powder format for long-term storage and easy reconstitution.