Learn More
Medchemexpress LLC Biotin alkyne | 773888-45-2 | 99.53% | 281.37 | 25 MG
SDP
Catalog No. 5000348462
Encompass_Preferred
Biotin alkyne is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. PROTACs themselves are compounds utilizing two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein, to exploit the intracellular ubiquitin-proteasome system for selective protein degradation. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-48462 Supplier Medchemexpress LLC Supplier No. 25MGHY138749
Biotin alkyne is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. PROTACs themselves are compounds utilizing two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein, to exploit the intracellular ubiquitin-proteasome system for selective protein degradation. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups