Learn More
Medchemexpress LLC Biotin alkyne | 773888-45-2 | MFCD28385487 | 98.6% | 281.37 | C13H19N3O2S | 5 MG
SDP
Catalog No. 5000222514
Encompass_Preferred
Biotin alkyne is a biotin-labeled, PEG-based linker containing a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is designed for introducing biotin tags in PROTAC synthesis, bioconjugation, and affinity capture workflows, and is supplied at high purity for research-scale applications.
  • Contains a terminal alkyne for CuAAC
  • Facilitates biotin labeling for affinity capture and detection
  • Peg-based linker provides solubility and flexibility
  • High purity suitable for synthesis and analytical use
  • Available in small quantities for research-scale workflows
Catalog No. 50-002-22514 Supplier Medchemexpress LLC Supplier No. HY1387495MG
Biotin alkyne is a biotin-labeled, PEG-based linker containing a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is designed for introducing biotin tags in PROTAC synthesis, bioconjugation, and affinity capture workflows, and is supplied at high purity for research-scale applications.
  • Contains a terminal alkyne for CuAAC
  • Facilitates biotin labeling for affinity capture and detection
  • Peg-based linker provides solubility and flexibility
  • High purity suitable for synthesis and analytical use
  • Available in small quantities for research-scale workflows