Learn More
TARGETMOL CHEMICALS INC Biotin-aniline 10MG
SDP
Catalog No. 502879990
Encompass_Preferred
Also available in 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Biotin-aniline is a probe that reacts strongly to RNA and DNA.Biotin-aniline is highly efficient and is used to capture subcellular transcriptomes in living cells with high spatial specificity. Purity 99.08%
Catalog No. 50-287-9990 Supplier TARGETMOL CHEMICALS INC Supplier No. T7207910MG
Also available in 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Biotin-aniline is a probe that reacts strongly to RNA and DNA.Biotin-aniline is highly efficient and is used to capture subcellular transcriptomes in living cells with high spatial specificity. Purity 99.08%