Learn More
Medchemexpress LLC HY-139912 50mg , Biotin-aniline CAS:769933-15-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13991250MG
Encompass_Preferred
HY-139912 50mg Biotin-aniline CAS: 769933-15-5 Biotin-aniline is a probe with substantially high reactivity towards RNA and DNA. Biotin-aniline emerges as more efficient probe for capturing subcellular transcriptome in living cells with high spatial specificity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1453