Learn More
Medchemexpress LLC Biotin-aniline | 769933-15-5 | 99.6% | 362.49 | 25 MG
SDP
Catalog No. 5000350904
Encompass_Preferred
Biotin-aniline is a probe that exhibits high reactivity towards RNA and DNA. It also functions as a novel APEX2 substrate and can label proteins via miniSOG. This product is noted for its efficiency in capturing subcellular transcriptome in living cells with high spatial specificity. Solutions should be aliquoted to prevent inactivation from repeated freeze-thaw cycles.
  • High reactivity towards RNA and DNA
  • Novel APEX2 substrate
  • Can label proteins via miniSOG
  • Efficient for capturing subcellular transcriptome in living cells
  • Provides high spatial specificity
Catalog No. 50-003-50904 Supplier Medchemexpress LLC Supplier No. 25MGHY139912
Biotin-aniline is a probe that exhibits high reactivity towards RNA and DNA. It also functions as a novel APEX2 substrate and can label proteins via miniSOG. This product is noted for its efficiency in capturing subcellular transcriptome in living cells with high spatial specificity. Solutions should be aliquoted to prevent inactivation from repeated freeze-thaw cycles.
  • High reactivity towards RNA and DNA
  • Novel APEX2 substrate
  • Can label proteins via miniSOG
  • Efficient for capturing subcellular transcriptome in living cells
  • Provides high spatial specificity