Learn More
Biolegend Biotin anti-mouse Ig light chain λ
Supplier:  Biolegend 407304
Encompass_Preferred
Biotin anti-mouse Ig light chain λ [RML-42]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ELISA, FC; Size: 500 ug
Catalog No. 50-169-197