Learn More
Biolegend Biotin anti-mouse Ig light chain κ
Supplier:  Biolegend 407204
Encompass_Preferred
Biotin anti-mouse Ig light chain κ [RMK-12]; Isotype: Rat IgG; Reactivity: Mouse; Apps: ELISA, FC; Size: 500 ug
Catalog No. 50-169-196