Learn More
Biolegend Biotin anti-Allophycocyanin (APC)
Supplier:  Biolegend 408004
Encompass_Preferred
Biotin anti-Allophycocyanin (APC) [APC003]; Isotype: Mouse IgG1, κ; Reactivity: 2nd Step; Apps: FC; Size: 100 ug
Catalog No. 50-207-9819