Learn More
Biolegend Biotin (azide-free) anti-β-Amyloid, 1-40
Supplier:  Biolegend 805404
Encompass_Preferred
Biotin (azide-free) anti-β-Amyloid, 1-40 [11A50-B10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA, WB, IHC, IP; Size: 200 uL
Catalog No. 50-207-3939