Learn More
ABclonal Technology Biotin anti- CD161c/NK1.1
SDP
Catalog No. 5000009881
Encompass_Preferred
Enables identical protein binding activity and signaling receptor activity Acts upstream of or within natural killer cell activation and positive regulation of natural killer cell mediated cytotoxicity Located in external side of plasma membrane Is integral component of plasma membrane Is expressed in thymus primordium Orthologous to human KLRB1 (killer cell lectin like receptor B1)
Catalog No. 50-000-09881 Supplier ABclonal Technology Supplier No. A27412200T
Enables identical protein binding activity and signaling receptor activity Acts upstream of or within natural killer cell activation and positive regulation of natural killer cell mediated cytotoxicity Located in external side of plasma membrane Is integral component of plasma membrane Is expressed in thymus primordium Orthologous to human KLRB1 (killer cell lectin like receptor B1)