Learn More
Biolegend Biotin anti-FITC
Supplier:  Biolegend 408304
Encompass_Preferred
Biotin anti-FITC [FIT-22]; Isotype: Mouse IgG1, λ; Reactivity: 2nd Step; Apps: FC; Size: 100 ug
Catalog No. 50-169-231