Learn More
Biolegend Biotin anti-human CD57
Supplier:  Biolegend 359616
Encompass_Preferred
Biotin anti-human CD57 [HNK-1]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-207-7872