Learn More
Biolegend Biotin anti-human CD117 (c-kit)
Supplier:  Biolegend 323406
Encompass_Preferred
Biotin anti-human CD117 (c-kit) [A3C6E2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-167-190