Learn More
Biolegend Biotin anti-human CD137 (4-1BB)
Supplier:  Biolegend 309806
Encompass_Preferred
Biotin anti-human CD137 (4-1BB) [4B4-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 ug
Catalog No. 50-166-525