Learn More
Biolegend Biotin anti-human CD163
Supplier:  Biolegend 333603
Encompass_Preferred
Biotin anti-human CD163 [GHI/61]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Cynomolgus, Rhesus; Apps: FC; Size: 25 ug
Catalog No. 50-167-725