Learn More
Biolegend Biotin anti-human CD19
Supplier:  Biolegend 302203
Encompass_Preferred
Biotin anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC; Size: 25 ug
Catalog No. 50-165-822