Learn More
Biolegend Biotin anti-human CD1c
Supplier:  Biolegend 331504
Encompass_Preferred
Biotin anti-human CD1c [L161]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 ug
Catalog No. 50-167-652