Learn More
Biolegend Biotin anti-human CD2
Supplier:  Biolegend 300203
Encompass_Preferred
Biotin anti-human CD2 [RPA-2.10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 25 ug
Catalog No. 50-165-516