Learn More
Biolegend Biotin anti-human CD20
Supplier:  Biolegend 302349
Encompass_Preferred
Biotin anti-human CD20 [2H7]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Squirrel Monkey; Apps: FC; Size: 25 ug
Catalog No. 50-207-2868