Learn More
Biolegend Biotin anti-human CD252 (OX40L)
Supplier:  Biolegend 326306
Encompass_Preferred
Biotin anti-human CD252 (OX40L) [11C3.1]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green Monkey; Apps: FC; Size: 100 ug
Catalog No. 50-167-319