Learn More
Biolegend Biotin anti-human CD26
Supplier:  Biolegend 302718
Encompass_Preferred
Biotin anti-human CD26 [BA5b]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-207-2898