Learn More
Biolegend Biotin anti-human CD275 (B7-H2, ICOSL)
Supplier:  Biolegend 309406
Encompass_Preferred
Biotin anti-human CD275 (B7-H2, ICOSL) [2D3]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-166-510