Learn More
Biolegend Biotin anti-human CD284 (TLR4)
Supplier:  Biolegend 312804
Encompass_Preferred
Biotin anti-human CD284 (TLR4) [HTA125]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Guinea Pig; Apps: FC; Size: 100 ug
Catalog No. 50-166-701