Learn More
Biolegend Biotin anti-human CD3
Supplier:  Biolegend 300403
Encompass_Preferred
Biotin anti-human CD3 [UCHT1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 ug
Catalog No. 50-165-554