Learn More
Biolegend Biotin anti-human CD3
Supplier:  Biolegend 344820
Encompass_Preferred
Biotin anti-human CD3 [SK7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, ICC; Size: 100 ug
Catalog No. 50-168-315