Learn More
Biolegend Biotin anti-human CD354 (TREM-1)
Supplier:  Biolegend 314904
Encompass_Preferred
Biotin anti-human CD354 (TREM-1) [TREM-26]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, ELISA Detection; Size: 100 ug
Catalog No. 50-166-820