Learn More
Biolegend Biotin anti-human CD45RO
Supplier:  Biolegend 304220
Encompass_Preferred
Biotin anti-human CD45RO [UCHL1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Chimpanzee, Cynomolgus, Common Marmoset; Apps: FC; Size: 100 ug
Catalog No. 50-166-104