Learn More
Biolegend Biotin anti-human IL-1β
Supplier:  Biolegend 511703
Encompass_Preferred
Biotin anti-human IL-1β [H1b-98]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Detection, ELISPOT Detection, ICC; Size: 50 ug
Catalog No. 50-170-101