Learn More
Biolegend Biotin anti-human IL-7
Supplier:  Biolegend 506601
Encompass_Preferred
Biotin anti-human IL-7 [BVD10-11C10]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ELISA Detection, ELISPOT Detection; Size: 50 ug
Catalog No. 50-169-982