Learn More
Biolegend Biotin anti-human/mouse CXCR7
Supplier:  Biolegend 331111
Encompass_Preferred
Biotin anti-human/mouse CXCR7 [8F11-M16]; Isotype: Mouse IgG2b, κ; Reactivity: human, mouse; Apps: FC, ICFC; Size: 25 ug
Catalog No. 50-167-615