Learn More
Biolegend Biotin anti-human/mouse integrin β7
Supplier:  Biolegend 321209
Encompass_Preferred
Biotin anti-human/mouse integrin β7 [FIB504]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Mouse, Rhesus; Apps: FC; Size: 50 ug
Catalog No. 50-167-081