Learn More
Biolegend Biotin anti-human TCR γ/δ
Supplier:  Biolegend 331206
Encompass_Preferred
Biotin anti-human TCR γ/δ [B1]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC, IHC; Size: 100 ug
Catalog No. 50-167-624