Learn More
Biolegend Biotin anti-human LT-α (TNF-β)
Supplier:  Biolegend 503104
Encompass_Preferred
Biotin anti-human LT-α (TNF-β) [359-81-11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Detection, ELISPOT Detection, ICFC; Size: 500 ug
Catalog No. 50-169-785