Learn More
Biolegend Biotin anti-human CD33
Supplier:  Biolegend 303426
Encompass_Preferred
Biotin anti-human CD33 [WM53]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee; Apps: FC; Size: 100 ug
Catalog No. NC1529394