Learn More
Biolegend Biotin anti-human CD159a (NKG2A)
Supplier:  Biolegend 375118
Encompass_Preferred
Biotin anti-human CD159a (NKG2A) [S19004C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-207-8832