Learn More
Cytek Biosciences Inc Antibodies Biotin Anti-Human CD20 (2H7) ..... Cat. No. 30-0209-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 300209U025
Encompass_Preferred
Antibodies Biotin Anti-Human CD20 (2H7) ..... Cat. No. 30-0209-U025 ..... Size 25 µgThe 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation.The 2H7 antibody is reported to be cross-reactive with several non-human species including Baboon, Chimpanzee, Cynomolgus and Rhesus. Please choose the appropriate format for each application.
Catalog No. 50-210-3157