Learn More
Cytek Biosciences Inc Antibodies Biotin Anti-Human CD45 (HI30) ..... Cat. No. 30-0459-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 300459U025
Encompass_Preferred
Antibodies Biotin Anti-Human CD45 (HI30) ..... Cat. No. 30-0459-U025 ..... Size 25 µgThe HI30 antibody reacts with human CD45, one of the most abundant hematopoietic markers and one that is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, CD45RO and others.The HI30 antibody is widely used as a marker for human CD45 expression on T cells, B cells, monocytes, macrophages, and NK cells.
Catalog No. 50-210-3168