Learn More
Biolegend Biotin anti-human CX3CR1
Supplier:  Biolegend 341617
Encompass_Preferred
Biotin anti-human CX3CR1 [2A9-1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 50 μg
Catalog No. 50-272-2256