Learn More
Biolegend Biotin anti-human MICA/MICB
Catalog No. 502722358
Encompass_Preferred
Biotin anti-human MICA/MICB [6D4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 μg
Catalog No. 50-272-2358 Supplier Biolegend Supplier No. 320904
Biotin anti-human MICA/MICB [6D4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 μg