Learn More
Cytek Biosciences Inc Antibodies Biotin Anti-Human/Mouse CD44 (IM7) ..... Cat. No. 30-0441-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 300441U025
Encompass_Preferred
Antibodies Biotin Anti-Human/Mouse CD44 (IM7) ..... Cat. No. 30-0441-U025 ..... Size 25 µgThe IM7 antibody recognizes CD44, a ubiquitously expressed cell surface receptor which is important for extracellular matrix organization, cell-cell and cell-matrix adhesion and migration. CD44 may be expressed in a number of different isoforms (splice variants) from the most typical or œstandard form, known as CD44s, to variants designated CD44v, e.g. CD44v1 or CD44v6. These receptors interact with several ligands, but most often associate with an extracellular matrix component hyaluronate, through which it mediates adhesion.The IM7 antibody may be used for detection of all isoforms of CD44, as it recognizes constant epitopes near the extracellular proximal domain. (Xu et al, 2002, J. Leukoc. Biol. 72:1133-1141). It has been reported to be cross-reactive with many non-human species including Baboon, Chimpanzee, Cynomolgus, Rhesus, Horse, Cow, Pig, Dog and Cat CD44.
Catalog No. 50-210-3163