Learn More
Biolegend Biotin anti-human CD63
Supplier:  Biolegend 353017
Encompass_Preferred
Biotin anti-human CD63 [H5C6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 50 ug
Catalog No. NC0523477